Muffle furnaces with fold down door, Tmax 1200 °C and controller C550, L 3/12 - L 40/12
Leverancier: Nabertherm
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Nabertherm
455-1559EA 6460 EUR
455-1559 455-1339 455-1338 455-1838 455-1839 455-1344 455-1027 455-1126 455-1840 455-1345 455-1744 455-1128 455-1029 455-1028 455-1347 455-1127 455-1548 455-1746 455-1845 455-1745 455-1549 455-1846 455-1848 455-1034 455-1551 455-1133 455-1751 455-1035 455-1134 455-1037 455-1752 455-1136 455-1557 455-1337 455-1754 455-1556
Muffle furnaces with fold down door, Tmax 1200 °C and controller C550, L 3/12 - L 40/12
Ovens Verassings- en moffelovens
The L 3/12 - L 40/12 muffle furnaces are the ideal choice for daily laboratory use. These high quality models have an advanced and attractive design, and are very reliable. The muffle furnaces have a fold down door.
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Fold down door (L models) can be used as a work platform
  • Exhaust air outlet on the rear wall of furnace
  • Solid state relays provide low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive
Heating is from two sides by means of ceramic heating plates (heating from three sides for muffle furnaces L 24/12 - L 40/12). These ceramic heating plates with an integral heating element are protected from fumes and splashing, and are easy to replace. Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used.
The furnace is equipped with C550 professional controller with touch screen display, featuring 10 freely adjustable programs and 20 segments and two extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The C550 controllers can automatically activate the vent with fan or catalyst (not model L 15).
Bestelinformatie: All units require a 230 V single-phase power supply, except L 24 and L 40 (400 V 3/N/PE). Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of the furnace. Manual or automatic gas supply systems are also available. Corresponding models with fold down/lift up doors, different Tmax and other controllers are also available. Please search the website for more details. Corresponding models with lift up doors, different Tmax and other controllers are also available. Please search the website for more details.
Opgelet: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.