Chemical resistant gloves, Tychem® NT420
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Tychem®
112-5472EA 0 EUR
112-5472 112-5473 112-5475
Chemical resistant gloves, Tychem® NT420
Handschoenen
These ambidextrous 100% nitrile gloves are very resistant and provide a barrier to most oil grease and hydrocarbons.
  • Easy to tear away in case of emergency break
  • Easy one-hand use
  • Second-skin feel significantly reduces hand fatigue
  • Natural rubber and powder-free
  • Smooth grip
  • Low modulus formulation
Perfect for jobs requiring chemical protection, while keeping tactile discrimination. Recommended for food contact, laboratory and small parts handling.
Certificaten: EN ISO 374-1:2016 Type B (JKOPT), EN ISO 374-5:2016 (BACTERIA & FUNGI); Cat. III
Food contact approved.