Netherlands
Order Entry
Industrial gloves, Camapur® Comfort 616
Industrial gloves, Camapur® Comfort 616
Supplier:  KCL by Honeywell
Certificates
undefined
Industrial gloves, Camapur® Comfort 616
Supplier:  KCL by Honeywell
Specifications
  • Catalog No:
  • KCHL616.8
  • KCHL616.7
  • KCHL616.11
  • KCHL616.10
  • KCHL616.6
  • KCHL616.9
  • Model:
  • Comfort 616
  • Comfort 616
  • Comfort 616
  • Comfort 616
  • Comfort 616
  • Comfort 616
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388 (3131)
  • EN 388 (3131)
  • EN 388 (3131)
  • EN 388 (3131)
  • EN 388 (3131)
  • EN 388 (3131)
Specifications
Product Information
  • Catalog #
    Supplier #
    Glove size
    Colour
    Length
    Thickness
    Pk
    Availability
    Price
  • KCHL616.6
    616.6
    6
    White
    220 - 260 mm
    1,0 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 86,60 €
    86,60 €
    1 * 10 PAAR
       86,60 €
    Supplier Article Number: 616.6
    Local Article Number: KCHL616.6
    Specifications:
    More
    Specifications
    Model
    Comfort 616
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Cuff style
    Knit wrist
    EN norm
    EN 388 (3131)
  • KCHL616.7
    616.7
    7
    White
    220 - 260 mm
    1,0 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 86,60 €
    86,60 €
    1 * 10 PAAR
       86,60 €
    Supplier Article Number: 616.7
    Local Article Number: KCHL616.7
    Specifications:
    More
    Specifications
    Model
    Comfort 616
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Cuff style
    Knit wrist
    EN norm
    EN 388 (3131)
  • KCHL616.8
    616.8
    8
    White
    220 - 260 mm
    1,0 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 86,60 €
    86,60 €
    1 * 10 PAAR
       86,60 €
    Supplier Article Number: 616.8
    Local Article Number: KCHL616.8
    Specifications:
    More
    Specifications
    Model
    Comfort 616
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Cuff style
    Knit wrist
    EN norm
    EN 388 (3131)
  • KCHL616.9
    616.9
    9
    White
    220 - 260 mm
    1,0 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 86,60 €
    86,60 €
    1 * 10 PAAR
       86,60 €
    Supplier Article Number: 616.9
    Local Article Number: KCHL616.9
    Specifications:
    More
    Specifications
    Model
    Comfort 616
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Cuff style
    Knit wrist
    EN norm
    EN 388 (3131)
  • KCHL616.10
    616.10
    10
    White
    220 - 260 mm
    1,0 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 86,60 €
    86,60 €
    1 * 10 PAAR
       86,60 €
    Supplier Article Number: 616.10
    Local Article Number: KCHL616.10
    Specifications:
    More
    Specifications
    Model
    Comfort 616
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Cuff style
    Knit wrist
    EN norm
    EN 388 (3131)
  • KCHL616.11
    616.11
    11
    White
    220 - 260 mm
    1,0 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 86,60 €
    86,60 €
    1 * 10 PAAR
       86,60 €
    Supplier Article Number: 616.11
    Local Article Number: KCHL616.11
    Specifications:
    More
    Specifications
    Model
    Comfort 616
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Cuff style
    Knit wrist
    EN norm
    EN 388 (3131)