D-(+)-Galactose ≥98%, Biotech Reagent, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

2102-07EA 0 EUR
2102-07
D-(+)-Galactose ≥98%, Biotech Reagent, J.T.Baker®
D-(+)-Galactose
Formule: C₆H₁₂O₆
MW: 180,16 g/mol
MDL Nummer: MFCD00151230
CAS nummer: 59-23-4
https://nl.vwr.com/stibo/web/std.lang.all/21/21/2302121.jpg

Order Now

Specification Test Results

GMP Manufactured Product
Arsenic (As) ≤0,500 ppm
Assay (by HPLC) ≥98%
Glucose ≤2,0%
Heavy Metals (as Cu) ≤10 ppm
Iron (Fe) ≤5 ppm
Endotoxin Concentration (EU/g) ≤10
Residual Methanol ≤3000 ppm
Residual Ethanol ≤5000 ppm
EP - Appearance of Solution Passes Test
EP - Acidity or Alkalinity Passes Test
EP - Identification A Passes Test
EP - Specific Optical Rotation [Å]²º^D (+) 78,0 - 81,5 Degree
EP - Barium (Ba) Passes Test
EP - Lead (Pb) ≤0,5 ppm
EP - Water (H₂O) ≤1,0%
EP - Ash (sulphated) ≤0,1%
EP - Total Viable Count (per g) ≤100
Only Class 2 (Methanol) and Class 3 (Ethanol) residual solvents are present. Both are tested, quantified, and reported.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR