Biological indicators, ampoules for steam sterilisation, STERIL CONTROL
Leverancier: Liofilchem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
91052-VWREA 250 EUR
91052-VWR 91040. 91050-VWR 91054-VWR
Biological indicators, ampoules for steam sterilisation, STERIL CONTROL
Sterilisatie-indicatoren
Biological indicators for validating sterilisation using steam sterilisation (autoclaves).
Ampoules with nutrient medium, pH indicator and spores of Geobacillus stearothermophilus (GST).