Microcentrifuge tubes with socket screw caps
Leverancier: VWR, part of Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
525-1234EA 88.7 EUR
525-1234 525-1244 525-0650 525-1233 525-1243 525-1231 525-1241 525-1240 525-0648 525-0646 525-1239 525-1238 525-0644 525-1237 525-1236 525-0642 525-1235 525-0652
Microcentrifuge tubes with socket screw caps
Buisjes Microcentrifugebuisjes
These clear, high grade PP tubes are ideal for sample processing applications.
  • RNase-, DNase- and endotoxin-free
  • Sterile tubes are sterilised by irradiation
Excellent for freezer storage or secure sample handling. Screw caps incorporate silicone O-rings for a leakproof seal and accept colour coded inserts. Inserts snap into the top of the cap to create a flush surface. Tubes are available with standard or loop caps, which tether the cap to the tube.
Caps and tubes are steam autoclavable at 121 °C and freezable to −80 °C.
Certificaten: Manufactured in a Class 100 000 cleanroom environment.
Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems.