This thermometer helps to monitor temperature and humidity conditions in labs, offices, and storage areas.
- Jumbo display
- Accurate, reliable measurements with included NIST-traceable calibration
- Perfect for both indoor and outdoor applications
- LCD (2-digit, 1,125" H for RH; 3-digit,1,125" H for temperature)
- Can be wall-mounted or used with the flip-open stand
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Certificaten: NIST-traceable calibration also helps meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Leveringsinformatie: Includes submersible temperature sensor on a 3 m cable, one AAA battery, and NIST-traceable calibration certificate with data.