Pesticide reference standards, Saflufenacil Metabolites, analytical standards, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38586-10MGEA 81.8 EUR
38586-10MG 18126-10MG 76646-10MG
Pesticide reference standards, Saflufenacil Metabolites, analytical standards, Supelco®
Standaarden Organische standaarden
Saflufenacil metabolite M800H11 may be used as a reference standard in the determination of saflufenacil metabolite M800H11 in maize using QuEChERS solid phase extraction technique with liquid chromatography coupled with tandem mass spectrometry (SPE-LC-MS/MS).