Netherlands
Order Entry
Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • HILB-5-1510
  • HILB-5-0510
  • HILB-5-2520
  • HILB-5-1010
  • HILB-5-3030
  • HILB-5-3020
  • HILB-5-2530
  • HILB-5-7530
  • HILB-5-7520
  • HILB-5-1530
  • HILB-5-1520
  • HILB-5-1220
  • HILB-5-1030
  • HILB-5-1020
  • HILB-5-0530
  • HILB-5-0520
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • 4%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Max. pressure:
  • 1000 bar
  • pH range:
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • Surface area:
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • USP code:
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Information
  • Catalog #
    Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • HILB-5-2520
    HILIC-B
    Semi-preparative
    Stainless steel
    100 Å
    5 µm
    21,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    1000 bar
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-1510
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • HILB-5-0510
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • HILB-5-1010
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • HILB-5-3030
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-3020
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-2530
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-7530
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-7520
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-1530
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-1520
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-1220
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-1030
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-1020
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-0530
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
  • HILB-5-0520
    HILIC-B
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Carbon load
    4%
    Endcapped
    No
    Max. pressure
    pH range
    2 - 7
    Surface area
    300 m²/g
    USP code
    L8
    Environmental certification
Frequently Bought Together