Cleanroom face mask with headband
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HOIT110193EA 543 EUR
HOIT110193
Cleanroom face mask with headband
Face masks with two/three layers.
  • Made of polycellulose
  • Round flad head mask
Good breathability and protection from contamination around 10⁹ resistance.
Size of the mask is 180/210 mm.
Verpakking: 50 pieces/bag, 20 bags/case.