Avantor® Hichrom, Titanium Frit, PEEK Encapsulated
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Avantor® Hichrom
HICHHI-680EA 0 EUR
HICHHI-680
Avantor® Hichrom, Titanium Frit, PEEK Encapsulated
Filters Voorkolom filters en frits
Titanium frits are resistant to corrosion, biocompatible and recommended for protein analysis.