Netherlands
Order Entry
VWR®, Fuel testing standards - jet aviation fuel
VWR®, Fuel testing standards - jet aviation fuel
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Fuel testing standards - jet aviation fuel
Supplier:  VWR Chemicals
Certificates
Product Information
  • Catalog #
    Concentration
    Temperature
    Test method
    Pack type
    Pk
    Availability
    Price
  • Acidity Standard, Jet Fuel
    Acidity Standard, Jet Fuel
    0,0083 mg KOH/g
    ASTM D3242
    Aluminium bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    1 * 250 mL
       292,00 €
    Local Article Number: VWRC99122.230
  • Aniline Point Standard, Jet Fuel
    Aniline Point Standard, Jet Fuel
    99123.230
    58,48 °C
    ASTM D611
    Aluminium bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    1 * 250 mL
       292,00 €
    Local Article Number: VWRC99123.230
  • FIA Aromatics & Saturates Standard, Jet Fuel
    FIA Aromatics & Saturates Standard, Jet Fuel
    17,7% (vol.)
    ASTM D1319
    Aluminium bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    1 * 250 mL
       292,00 €
    Local Article Number: VWRC99124.230
  • Sulphur Standard, Jet Fuel
    Sulphur Standard, Jet Fuel
    99126.230
    7,4 mg/kg
    ASTM D3227 / IP 342 / ISO 3012
    Aluminium bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    1 * 250 mL
       292,00 €
    Local Article Number: VWRC99126.230
  • Freezing Point Standard, Jet Fuel
    Freezing Point Standard, Jet Fuel
    –55,5 °C
    ASTM D2386, IP 16, ISO 3013
    Aluminium bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    1 * 250 mL
       292,00 €
    Local Article Number: VWRC99127.230