U heeft gezocht op: Kleefmatten
16  results were found
SearchResultCount:"16"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: VWR Collection
Omschrijving: 30- of 60-laags zelfklevende entreematten vervaardigd uit polyethyleen met lage dichtheid. Deze matten houden vervuiling onder controle door vuil van ...
Leverancier: NEW PIG
Omschrijving: Provide a no-slip path in hazardous walking areas with our super-safe, adhesive-backed absorbent runner - its hi-viz yellow safety edges can't be miss...
Leverancier: JUSTRITE MANUFACTURING CO
Omschrijving: Diamond Statâ„¢ is a dissipative/anti-static mat designed to absorb static electricity.
Leverancier: NEW PIG
Omschrijving: Keep your restrooms cleaner and provide better drip and splash protection with these adhesive-back urinal mats.
Leverancier: JUSTRITE MANUFACTURING CO
Omschrijving: Anti-Stat P.O.P.â„¢ is a vinyl work surface that dissipates static from conductive items placed on it.
Leverancier: 3M
Omschrijving: 4300 matten voor verontreinigingsbeheersing bestaan uit 40 lagen met lijm beklede, doorzichtige polyethyleenvellen bevestigd aan een vinylrug. Voor ge...
Leverancier: NEW PIG
Omschrijving: Prevent slips, trips and falls in high traffic areas with our easy to clean, adhesive backed commercial floor mat.
Leverancier: Texwipe
Omschrijving: Meerlaagse kleefmat, CleanStep, 60 lagen, grijs, l×b: 457×1168 mm
Leverancier: NEW PIG
Omschrijving: These adhesive-backed toilet mats absorb drips and splashes of water or urine to keep the floor around toilet bases dry and odor free.
Catalogus nummer: (115-2586)
Leverancier: NEW PIG
Omschrijving: Adhesive-backed mat runner lets you create custom-sized restroom mats for urinals, sinks and other wet spots.
UOM: 1 * 1 ROL
Catalogus nummer: (115-2579)
Leverancier: NEW PIG
Omschrijving: Adhesive-backed mat bundle has everything you need to keep the floor in a small restroom clean, dry and safe, Includes urinal, toilet, plunger, and si...
UOM: 1 * 5 ST
Leverancier: NEW PIG
Omschrijving: Keep restroom floors dry and safe when people wash their hands with our adhesive-backed mats. Absorbent top soaks up water and other liquids.
Catalogus nummer: (115-2587)
Leverancier: NEW PIG
Omschrijving: Adhesive-backed mats keep the surfaces under plungers, brushes and soap dispensers neat and clean. Absorbent top soaks up water, urine and other liqui...
UOM: 1 * 6 ST
Catalogus nummer: (115-2469)
Leverancier: ERGOMAT
Omschrijving: Ergomat Sticky Mats provide a fast, convenient, reliable way to clean shoe soles, or other items, before entering a clean and sanitary area. They sign...
UOM: 1 * 10 ST
Leverancier: Nitritex
Omschrijving: Supatacâ„¢ Access Mats are made from LDPE with Polyacrylate adhesive and consist of 30 sheets. They prevent contamination and particulate matter from en...
Leverancier: PURUS INTERNATIONAL, INC.
Omschrijving: A choice of either 30 or 60 layer adhesive entrance mats are manufactured from low density polyethylene. The mats control contamination by removing de...
Sale
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 16
no targeter for Bottom