U heeft gezocht op: Canules
29  results were found
SearchResultCount:"29"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (BDAM367286)
Leverancier: Vyaire
Omschrijving: Blood collection sets consist of a needle and a multi-sample Luer adapter, connected by several length of tubing. A Safety-Lok protective shield reduc...
UOM: 1 * 50 ST
Leverancier: KIMA SAS DI CHIARIN RENZO & C
Omschrijving: Sterile needles with safety device for blood collection.
Leverancier: BD Medical
Omschrijving: Peripheral IV cannula with injection port and obturator.
Leverancier: BD Medical
Omschrijving: Deze canules zijn gemaakt van radiopaak FEP-polymeer en thermisch gevormde afschuiningen.
Leverancier: BD Medical
Omschrijving: These IV cannulas with polyurethane catheter and injection port are designed to access the peripheral veins of the patient’s blood system for re-hydra...
Leverancier: BD Medical
Omschrijving: Split-septum-apparaat elimineert de complexiteit van mechanische kleppen en daarmee bacteriën. Door het rechte en vrije pad garandeert het:
New Product
Leverancier: BD Medical
Omschrijving: Cannula, BD Insyte™ Autoguard™, Meter: G 20, roos, Ø×l: 1,1×30 mm
Catalogus nummer: (BRAU4190050)
Leverancier: B. Braun
Omschrijving: Aeration cannula maintains a bacteria-retentive venting of noncollapsing plastic containers.
UOM: 1 * 100 ST
Leverancier: B. Braun
Omschrijving: Vasofix Certo IV catheter with injection port. IV catheter material available in PU and FEP for low-pain, gentle insertion of catheter. Integrated inj...
Catalogus nummer: (BRAU4512383)
Leverancier: B. Braun
Omschrijving: Epidurale anesthesienaald met schuine kant van het Tuohy-type.
UOM: 1 * 100 ST
Catalogus nummer: (BRAU4439848)
Leverancier: B. Braun
Omschrijving: Rechte canules voor kortdurende infusies, bolusinjecties en poortirrigatie.
UOM: 1 * 50 ST
Leverancier: B. Braun
Omschrijving: Introcan Safety passief afgeschermde canules met zelfactiverende bescherming tegen prikaccidenten en gerelateerde infecties.
Catalogus nummer: (BRAU4056310N)
Leverancier: B. Braun
Omschrijving: Venofix sets with puncture needle made of high-quality chromium-nickel steel and a micro-siliconised precision bevel.
UOM: 1 * 50 ST
Catalogus nummer: (SOCO370.30500)
Leverancier: SOCOREX ISBA
Omschrijving: This spray nozzle cannula fits on Luer-Lock self-refilling syringes.
UOM: 1 * 1 ST
Leverancier: SOCOREX ISBA
Omschrijving: Blunt end cannulas provide precise distribution with Luer nozzle microdispensers.
Catalogus nummer: (WHEA851412)
Leverancier: DWK Life Sciences
Omschrijving: Blunt end dispensing cannulas with Luer Lock. Ideal for use with 24 and 96 well microplates.
UOM: 1 * 1 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 29
no targeter for Bottom