U heeft gezocht op: Diagnostische systemen
169  results were found
SearchResultCount:"169"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (COVID19AGVCDS)
Leverancier: SURESCREEN
Omschrijving: The COVID-19 Antigen Rapid Test Cassette is a rapid chromatographic immunoassay for the qualitative detection of COVID-19 antigen in saliva. The ident...
UOM: 1 * 1 EA
Catalogus nummer: (720-1350)
Leverancier: SURESCREEN
Omschrijving: Test kits for the swift diagnosis of infectious diseases.
UOM: 1 * 40 ST
Leverancier: GOLD STANDARD DIAGNOSTICS
Omschrijving: This ELISA kit is intended for the qualitative detection of IgA class antibodies to SARS-CoV-2 virus in human serum and plasma for diagnostics of expo...
Leverancier: GOLD STANDARD DIAGNOSTICS
Omschrijving: This RT-PCR assay is a highly sensitive Multiplex Real-Time PCR with CE-IVD marking for the direct qualitative pathogen detection of SARS-CoV-2 genomi...
Catalogus nummer: (GSDCCVAG4080A)
Leverancier: GOLD STANDARD DIAGNOSTICS
Omschrijving: GSD NovaGen SARS-CoV-2 Ag rapid test completes the diagnostic panel of the Gold Standard Diagnostics Group for SARS-CoV-2 diagnostics: ELISA, RT-PCR, ...
UOM: 1 * 1 EA
Leverancier: SURESCREEN
Omschrijving: To determine health function from a urine sample. The Urinalanalysis test strip fetures various colour change pads that indicate correct concentration...
Catalogus nummer: (720-1356)
Leverancier: SURESCREEN
Omschrijving: This test detects the cause of syphilis, Treponema Pallidum (TP).
UOM: 1 * 1.000 ST
Leverancier: SURESCREEN
Leverancier: Vyaire
Omschrijving: Blood Collection tube, BD Vacutainer®, BD Vacutainer®, Glas, Diameter: 16 mm, Lengte: 100 mm
Catalogus nummer: (KNAUA0006AC)
Leverancier: KNAUER
Omschrijving: The K-7400S semi-mirco osmometer enables the measurement of osmolality as well as the freezing point of aqueous solutions. The proven technology of fr...
UOM: 1 * 1 ST
Leverancier: Vyaire
Omschrijving: PET, clear.
Leverancier: GOLD STANDARD DIAGNOSTICS
Omschrijving: Real-Time RT-PCR kit for the qualitative detection and discrimination of relevant mutations associated with SARS-CoV-2 variants: B.1.1.7 (UK), B.1.351...
Catalogus nummer: (720-1345)
Leverancier: SURESCREEN
Omschrijving: Fecal occult blood (FOB) test are rapid tests for the detection of faecal occult blood.
UOM: 1 * 25 ST
Catalogus nummer: (SARSSBK25)
Leverancier: BIOMIRE
Omschrijving: The COV-Hygien Xpress detection kit provides rapid, ready to use technology to detect SARS CoV-2 on surfaces and liquids.
UOM: 1 * 1 EA
Leverancier: GOLD STANDARD DIAGNOSTICS
Omschrijving: This Real-Time RT-PCR assay is intended for the qualitative detection of SARS-CoV-2 from human nasopharyngeal swabs, eluted directly into deionised wa...
Catalogus nummer: (GSDCCVGQ0970)
Leverancier: GOLD STANDARD DIAGNOSTICS
Omschrijving: Immunoassay for the quantitative and qualitative detection of specific IgG-class antibodies to the spike protein of SARS-CoV-2.
UOM: 1 * 1 EA
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 169
no targeter for Bottom