U heeft gezocht op: Infusen
27  results were found
SearchResultCount:"27"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (BRAU4099303)
Leverancier: B. Braun
Omschrijving: Sterifix infusion filter is specially designed for bacteria and particle retention.
UOM: 1 * 50 ST
Leverancier: B. Braun
Omschrijving: Original-Perfusor tubings for use with all syringe pumps inferior to four-bar pressure.
Leverancier: B. Braun
Omschrijving: Suitable for all pressure infusion systems <2 bar.
Catalogus nummer: (BRAU16615C)
Leverancier: B. Braun
Omschrijving: Drug-Resistant Multi-Tap Systems are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, i...
UOM: 1 * 100 ST
Catalogus nummer: (BRAU415111)
Leverancier: B. Braun
Omschrijving: Simplify IV technique by direct insertion into a closed system.
UOM: 1 * 100 ST
Leverancier: B. Braun
Omschrijving: Heidelberger extension tubing for infusion systems.
Leverancier: B. Braun
Omschrijving: Convenient drip chamber design with sharp spike ensures easy piercing of container ports, bacteria proof venting (closable), 20 drops 1 ml ±0,1 ml (aq...
Catalogus nummer: (BRAU4099842N)
Leverancier: B. Braun
Omschrijving: Ready-to-use set with infusion filter.
UOM: 1 * 20 ST
Leverancier: B. Braun
Omschrijving: Sterile stopcock with rotating swivel nut.
Catalogus nummer: (BRAU4093054)
Leverancier: B. Braun
Omschrijving: Extension and connection tubing with small bore.
UOM: 1 * 100 ST
Catalogus nummer: (BRAU4099257)
Leverancier: B. Braun
Omschrijving: Infusion filter with bacteria and particle retention.
UOM: 1 * 1 ST
Catalogus nummer: (720-2571)
Leverancier: B. Braun
Omschrijving: Transferspike voor steriele vloeistoffen voor flessen tot 100 ml
UOM: 1 * 50 ST
Catalogus nummer: (BRAU4183916)
Leverancier: B. Braun
Omschrijving: Intrapur plus offers reliable air venting regardless of position with connection lines at both sides of the filter with Luer lock fittings.
UOM: 1 * 50 ST
Catalogus nummer: (BRAU16609C)
Leverancier: B. Braun
Omschrijving: Five-fold tap banks are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppress...
UOM: 1 * 40 ST
Catalogus nummer: (720-2580)
Leverancier: B. Braun
Omschrijving: Sterifix Paed is speciaal ontworpen voor het vasthouden van bacteriën en deeltjes.
UOM: 1 * 50 ST
Catalogus nummer: (BRAU409100H)
Leverancier: B. Braun
Omschrijving: This capless valve allows for a safe and convenient access during infusion therapy.
UOM: 1 * 50 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 27
no targeter for Bottom