U heeft gezocht op: Smeermiddelen
41  results were found
SearchResultCount:"41"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (1.07746.0100)
Leverancier: Merck
Omschrijving: Grease, silicone, density: 1 g/cm3 (20 °C), flash point >300 °C
UOM: 1 * 100 g
MSDS
Catalogus nummer: (6674.0050)
Leverancier: VWR Chemicals
Omschrijving: VWR® Chemicals silicone grease is a valve lubricant and sealant, often times used as a vacuum grease. It has good resistance to most chemicals, a wide...
UOM: 1 * 50 g
Catalogus nummer: (9930.5000)
Leverancier: VWR Chemicals
Omschrijving: VWR® TECHNICAL Vacuum oil for pump
UOM: 1 * 5 L
Leverancier: IKA
Omschrijving: These certified standard silicone oils can be used to check the measurement accuracy of ROTAVISC.
Catalogus nummer: (44224.KT)
Leverancier: Thermo Scientific
Omschrijving: High-vacuum grease is used as an anti-seize grease in ground glass joints and to lubricate synthetic and silicone rubber gaskets. Also used to lubrica...
UOM: 1 * 1 ST
MSDS
Catalogus nummer: (HETT4051)
Leverancier: Hettich
Omschrijving: Lubricating grease for rotor trunnions
UOM: 1 * 1 ST
MSDS
Catalogus nummer: (APOSPC32832-1PK)
Leverancier: Apollo Scientific
Omschrijving: Silica thickened inert grease.
UOM: 1 * 1 EA
Catalogus nummer: (BRDY834194)
Leverancier: Brady
Omschrijving: Non-greasing maintenance spray for lock cylinders, office machines, tools, car locks.
UOM: 1 * 1 ST
Catalogus nummer: (ACRO386110010)
Leverancier: Thermo Scientific
Omschrijving: Synthetic grease for joints.
UOM: 1 * 1 ST
MSDS
Catalogus nummer: (APPC211757.1211)
Leverancier: PanReac AppliChem
Omschrijving: It is a purified mixture of semisolid saturated hydrocarbons having the general formula CnH2n+2 and is obtained from petroleum.
UOM: 1 * 1.000 g
Leverancier: Welch by Gardner Denver
Omschrijving: Vacuum pump parafin oils, 10 l, 10⁻⁸ vapour pressure mbar, viscosity at 40 °C cSt 94, Flash point °C 260, density at 15 °C 0,896, LABOVAC 12S
Leverancier: VACUUBRAND GMBH + CO
Omschrijving: Rotatiepompolie B
Rotatiepompolie B heeft uitstekende viscositeitskenmerken en een lage dampdruk. Door de goede chemische resistentie, de ...
Leverancier: Welch by Gardner Denver
Omschrijving: A specially fractionated oil for DuoSeal® belt-driven pumps is designed to ensure the highest vacuum performance. The oil is tested to high vacuum lev...
Catalogus nummer: (24490.130)
Leverancier: M & I MATERIALS LTD
Omschrijving: Medium temperature vacuum grease. Silicone-free. Used when a grease of higher melting point is required.
UOM: 1 * 25 g
MSDS
Leverancier: M & I MATERIALS LTD
Omschrijving: These Apiezon greases are hydrocarbon based, silicone-free and highly resistant to creep, the undesirable phenomenon associated with silicone-based pr...
Catalogus nummer: (24492.136)
Leverancier: M & I MATERIALS LTD
Omschrijving: Apiezon N Grease exhibits extremely low vapour pressures at ambient temperatures, 6×10-10 torr at 20 °C, which are further improved by redu...
UOM: 1 * 25 g
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 41
no targeter for Bottom