U heeft gezocht op: Reactievaten, cilindrische vorm
21  results were found
SearchResultCount:"21"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: Helder Duran®-borosilicaatglas, duidelijk, met vlakke bodem.
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: Reaction vessel, made of DURAN® borosilicate glass, flat bottom, with easily readable graduation and flat laboratory flange (DN) with groove. With two...
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: Helder DURAN®-borosilicaatglas.
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: Reaction vessel, made of DURAN® borosilicate glass, round bottom, with easily readable graduation and laboratory flat flange (LF) with groove.
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: Helder DURAN® -borosilicaatglas, met ronde bodem.
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: DURAN® borosilicate glass.
Catalogus nummer: (PALL97134)
Leverancier: Cytiva (Formerly Pall Lab)
Omschrijving: Reactievaten, cilindrisch, Stainless steel pressure vessel, Roestvrij staal, Inhoud: 18,9 L (5 gallon)
UOM: 1 * 1 ST
Leverancier: KGW (KARLSRUHER GLASTECHN.)
Omschrijving: Borosilicate glass.
Catalogus nummer: (ROBU18910)
Leverancier: ROBU GLASFILTER
Omschrijving: CSB reaction vessel with moulded diameter and standard joint 29/32.
UOM: 1 * 2 ST
Leverancier: KGW (KARLSRUHER GLASTECHN.)
Omschrijving: Borosilicate glass.
Leverancier: KGW (KARLSRUHER GLASTECHN.)
Omschrijving: Reaction vessel with stationary frame, J valve and accessories
Leverancier: DWK Life Sciences
Omschrijving: DURAN®, borosilicaatglas 3.3, helder, met ronde bodem.
Catalogus nummer: (KGWATRGN7577)
Leverancier: KGW (KARLSRUHER GLASTECHN.)
Omschrijving: This reaction vessel is made of borosilicate glass 3.3
UOM: 1 * 1 ST
Catalogus nummer: (KGWATRGN7399)
Leverancier: KGW (KARLSRUHER GLASTECHN.)
Omschrijving: This cylindrical reaction vessel is made up of borosilicate glass.
UOM: 1 * 1 ST
Leverancier: DWK Life Sciences
Omschrijving: DURAN®, borosilicaatglas 3.3, helder.
Catalogus nummer: (WITG0.900.100)
Leverancier: WITEG LABORTECHNIK GMBH
Omschrijving: Borosilicate glass.
UOM: 1 * 1 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 21
no targeter for Bottom