Tritirachium album Proteinase K
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
390972XEA 159 EUR
VWRC390972X
Tritirachium album Proteinase K
Activity approximately 30 Anson units per g.
1 Anson unit is the amount of enzyme which liberates 1 mmol of Folin positive amino acids per minute at pH 7,5 and 35 °C using haemoglobin as substrate. Powerful proteolytic activity on proteins, glycoproteins and peptides. Used to inactivate DNases and RNases during isolation of nucleic acids.