Digoxigenin-dUTP, alkali stable
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40078.EA 321 EUR
40078.
Digoxigenin-dUTP, alkali stable
Reagentia voor nucleïnezuren Nucleotiden
Digoxigenin-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3'-end terminal labeling to synthesize labeled DNA probes for in-situ hybridization, microarray or blotting techniques. The digoxigenin (DIG) labeled probe could be detected using a labeled anti-digoxigenin antibody.