VWR® AVS TITRINORM, Barium Standard Solution, 1000 mg/L Ba in dil. Nitric Acid, Standard for AAS
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® AVS TITRINORM
86664.180EA 30.3 EUR
86664.180 86664.260
VWR® AVS TITRINORM, Barium Standard Solution, 1000 mg/L Ba in dil. Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Barium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022