Netherlands
Order Entry
VWR® ARISTAR®, Neodymium Standard Solution, 10000 mg/L Nd in dil. Nitric Acid (from Nd₂O₃), Standard for ICP
VWR® ARISTAR®, Neodymium Standard Solution, 10000 mg/L Nd in dil. Nitric Acid (from Nd₂O₃), Standard for ICP
Supplier:  VWR Chemicals
Certificates
undefined
VWR® ARISTAR®, Neodymium Standard Solution, 10000 mg/L Nd in dil. Nitric Acid (from Nd₂O₃), Standard for ICP
Supplier:  VWR Chemicals
Certificates
Related Information
Product Information
  • Catalog #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Neodymium
    Nd
    10000mg/L
    2-5% HNO₃
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1 250,00 €
    1 250,00 €
    1 * 500 mL
       1 250,00 €
  • 455712C
    Neodymium
    Nd
    10000mg/L
    2-5% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 387,00 €
    387,00 €
    1 * 100 mL
       387,00 €
Frequently Bought Together