VWR® ARISTAR®, Phosphorus Standard Solution, 10000 mg/L P in Water (from (NH₄)PO₄H₂), Standard for ICP
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® ARISTAR®
455794UEA 578 EUR
455794U 455792S
VWR® ARISTAR®, Phosphorus Standard Solution, 10000 mg/L P in Water (from (NH₄)PO₄H₂), Standard for ICP
Standaarden Kationische standaarden Fosfor standaarden
NH₄H₂PO₄ in H₂O.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022