Netherlands
Order Entry
Selenium standard solution, 1000 mg/L Se in 0,5 M nitric acid (from SeO₂) Certipur® standard for AAS, Supelco®
Selenium standard solution, 1000 mg/L Se in 0,5 M nitric acid (from SeO₂) Certipur® standard for AAS, Supelco®
Supplier:  Merck
Certificates
Selenium standard solution, 1000 mg/L Se in 0,5 M nitric acid (from SeO₂) Certipur® standard for AAS, Supelco®
Supplier:  Merck
Certificates
Specifications
  • M-Clarity quality level:
    MQ100
Specifications
Related Information
Certificates
Search by Lot Number
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • 1.19796.0500
    1.19796.0500
    Selenium
    Se
    1000 mg/L
    0,5 M HNO₃
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 56,90 €
    56,90 €
    1 * 500 mL
       56,90 €
    Supplier Article Number: 1.19796.0500
    Local Article Number: MERC1.19796.0500
  • 1.19796.0100
    1.19796.0100
    Selenium
    Se
    1000 mg/L
    0,5 M HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 40,60 €
    40,60 €
    1 * 100 mL
       40,60 €
    Supplier Article Number: 1.19796.0100
    Local Article Number: MERC1.19796.0100