


Chloromethane (methyl chloride) ∼5.7% ∼1 mol/L in THF
TCIAM2813-100ML
: TCI
:
- Pk:100 mL
- Seq:2440
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (HNMR):5.5 to 6.0 w/w%
:
:14,06041
:3,II
:50.49 g/mol
:200-817-4
:1993
:GHS02
:GHS07
:GHS08
:Refrigerator
:74-87-3
:CH₃Cl
:MFCD00000872