Pregnancy test strips
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-0723EA 107 EUR
720-0723
Pregnancy test strips
Diagnostische systemen Clinical Reagents and Kits
An extremely accurate pregnancy test for early diagnosis in a low cost strip format.
  • Easy to use, and low cost, simply dip the strip into urine
  • More than 99% accurate
  • Ability to detect hCG, the chemical marker for pregnancy, as early as one day after the first missed menstrual period
  • Sensitive, with the ability to detect as low as 25 mIU/mL hCG in urine and/or serum specimens
  • Simple testing procedure; no special reagents or equipment required
  • Accurate, easy-to-read results in just minutes
Human chorionic gonadotropin, or hCG, is a hormone produced by the placenta shortly after the implantation of a fertilized egg into the womb.
The early appearance of hCG in both the urine and serum of pregnant women and its rapid rise in concentration during gestational growth make it an excellent marker for the early detection of pregnancy.
Certificaten: CE marked, FDA approved.