Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Supplier: Avantor
Certificates
About this item
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI66-25-015-50
- HI66-25-015-60
- HI66-25-025-15
- HI66-25-025-50
- HI78-10-010-5
- HI66-25-025-60
- HI66-25-025-25
- HI66-25-025-30
- HI78-10-010-10
- HI66-25-015-25
- HI66-25-015-30
- HI66-25-015-15
- Phase name:
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- Phase composition:
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Polarity:
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Crossbond:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Temperature limit:
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
Specifications
Product Information
- Catalog #Column FormatFilm thicknessØ int.LengthPkAvailabilityPrice
- Chiral GC columnChiral GC columnHI66-25-015-50Capillary0,15 µm0,25 mm50 m1Log in for availability1 * 1 ST2 100,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-015-60Capillary0,15 µm0,25 mm60 m1Log in for availability1 * 1 ST2 120,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnSpecifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-025-50Capillary0,25 µm0,25 mm50 m1Log in for availability1 * 1 ST2 100,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral FAST GC columnChiral FAST GC columnHI78-10-010-5Capillary0,10 µm0,10 mm5 m1Log in for availability1 * 1 ST873,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-025-60Capillary0,25 µm0,25 mm60 m1Log in for availability1 * 1 ST2 120,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-025-25Capillary0,25 µm0,25 mm25 m1Log in for availability1 * 1 ST1 090,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-025-30Capillary0,25 µm0,25 mm30 m1Log in for availability1 * 1 ST1 160,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral FAST GC columnChiral FAST GC columnHI78-10-010-10Capillary0,10 µm0,10 mm10 m1Log in for availability1 * 1 ST1 130,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-015-25Capillary0,15 µm0,25 mm25 m1Log in for availability1 * 1 ST1 090,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnChiral GC columnHI66-25-015-30Capillary0,15 µm0,25 mm30 m1Log in for availability1 * 1 ST1 160,00 €Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C - Chiral GC columnSpecifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-DEX DAC BetaPhase compositionDiacethyl TBS Beta Cyclodexstrin basedPolarityChiral stationary phaseCrossbondNoTemperature limit230 °C