Netherlands
Order Entry
Selenium AAS Standards, (CRMs), Fluka™
Selenium AAS Standards, (CRMs), Fluka™
Certificates
Selenium AAS Standards, (CRMs), Fluka™
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • HONCE4067-100ML
    E4067-100ML
    Selenium
    Se
    1000 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 34,70 €
    34,70 €
    1 * 100 mL
       34,70 €
    Supplier Article Number: E4067-100ML
    Local Article Number: HONCE4067-100ML
  • HONCE4067-500ML
    E4067-500ML
    Selenium
    Se
    1000 mg/L
    2% HNO₃
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 56,70 €
    56,70 €
    1 * 500 mL
       56,70 €
    Supplier Article Number: E4067-500ML
    Local Article Number: HONCE4067-500ML
  • HONCE4118-100ML
    E4118-100ML
    Selenium
    Se
    10000 mg/L
    5% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 139,00 €
    139,00 €
    1 * 100 mL
       139,00 €
    Supplier Article Number: E4118-100ML
    Local Article Number: HONCE4118-100ML