Knee pad, non-marking, KP50
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRTWKP50WHREA 16.6 EUR
PRTWKP50WHR
Knee pad, non-marking, KP50
Ergonomische steunen en hulpmiddelen Kniebeschermers
This knee pad made of EPE foam, polyester provides excellent protection and stability on all floor types.
  • Hard wearing outer shell for protection
  • Lightweight wrap-around design
  • Breathable inner fabric allows air to circulate
  • A thick foam centre cushions the wearers knee
  • Retail bag which aids presentation for retail sales
The moulded non-marking materials prevent marking and scratching to the work surface. Ergonomic design provides all round protection and comfort with superb grip to avoid sliding. Adjustable straps for secure and flexible fit at all times.
Certificaten: EN 14404:2004+A1:2010 Type 1 Level 1