VWR® Advanced, Verwarmingsplaten
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
444-0579EA 424 EUR
444-0579 VWRU97042-660 444-0576 444-0587 444-0586 444-0575 444-0589 444-0578 444-0577 444-0588 444-0583 444-0582 444-0585 VWRU97042-624 444-0584 444-0590 444-0581 444-0592 444-0591 444-0580
VWR® Advanced, Verwarmingsplaten
Verwarmers Verwarmingsplaten
VWR® Advanced hotplates deliver accurate and repeatable results. Units are microprocessor controlled and have an easy to read LED display for set point temperature. The control panel features easy to use controls which allow users to dial in temperature adjustments. Rear housing features an off-centred, integral support rod holder with locking knob to accept the optional support rod and clamp kit. The low profile design takes up less space and fits into fume hoods. A spill resistant housing channels fluids away from internal components.
  • Uitstekende temperatuuruniformiteit
  • Microprocessorregeling met ingebouwd geheugen voor het oproepen van de laatst ingestelde temperatuur (zelfs als het apparaat was uitgeschakeld)
  • Chemisch bestendige behuizing die koel aanvoelt
  • LED-display voor temperatuur
  • Keuze uit modellen met keramische of aluminium bovenplaten
A 'hot' symbol warning light is illuminated when heat is turned on and remains on until top plate cools down. Enhanced electronics regulate heating and bring samples to temperature quickly and efficiently. Temperature measurement is controlled more accurately by using a resistance thermometer (RTD). Ceramic tops feature a chemically resistant, reflective white top plate surface that is easy to clean. Durable aluminium tops will not crack or chip, and provide a more even heating surface.
Bestelinformatie: Er is een optionele kit met steunstaaf verkrijgbaar, inclusief roestvrijstalen steunstaaf van 457 mm, schuifklem voor thermometer/temperatuursonde, medium zwenkklem met 3 tanden, en bevestigingshaak.
Opgelet: * RT= Ambient.