Zwavelzuur 96%, CMOS for microelectronic, J.T.Baker®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
9674-37EA
0
EUR
9674-37
Zwavelzuur 96%, CMOS for microelectronic, J.T.Baker®
Zwavelzuur
Formule:
H₂SO₄ MW: 98,08 g/mol Kookpunt: ∼335 °C (1013 hPa) Smeltpunt: 3 °C Dichtheid: 1,84 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00064589 CAS nummer: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II |
Specification Test Results
Assay | 95.0 - 97.0 % |
Chloride (Cl) | max. 0.1 ppm |
Color (APHA) | max. 10 |
Heavy Metals (as Pb) | max. 200 ppb |
Nitrate (NO3) | max. 0.2 ppm |
Particles > 0.5 µm/ml | max. 150 |
Particles > 1.0 µm/ml | max. 25 |
Phosphate (PO4) | max. 0.3 ppm |
Residue after Ignition | max. 2 ppm |
Trace Impurities (in ppb) | 0 |
Aluminium (Al) | max. 50 |
Arsenic and Antimony (as As) | max. 5 |
Barium (Ba) | max. 10 |
Beryllium (Be) | max. 10 |
Bismuth (Bi) | max. 20 |
Boron (B) | max. 10 |
Cadmium (Cd) | max. 10 |
Calcium (Ca) | max. 50 |
Chromium (Cr) | max. 50 |
Cobalt (Co) | max. 10 |
Copper (Cu) | max. 10 |
Gallium (Ga) | max. 10 |
Germanium (Ge) | max. 10 |
Gold (Au) | max. 20 |
Iron (Fe) | max. 100 |
Lead (Pb) | max. 20 |
Lithium (Li) | max. 10 |
Magnesium (Mg) | max. 50 |
Manganese (Mn) | max. 10 |
Mercury (Hg) | max. 5 |
Molybdenum (Mo) | max. 10 |
Nickel (Ni) | max. 10 |
Niobium (Nb) | max. 10 |
Potassium (K) | max. 50 |
Silicon (Si) | max. 50 |
Silver (Ag) | max. 10 |
Sodium (Na) | max. 100 |
Strontium (Sr) | max. 10 |
Tantalum (Ta) | max. 10 |
Thallium (Tl) | max. 20 |
Tin (Sn) | max. 50 |
Titanium (Ti) | max. 10 |
Vanadium (V) | max. 10 |
Zinc (Zn) | max. 50 |
Zirconium (Zr) | max. 10 |
For Laboratory Research or Manufacturing use. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...