Autoradiografiecassettes, Amersham Hypercassette™
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Amersham™ Hypercassette™
RPN11642EA 301 EUR
RPN11642
Autoradiografiecassettes, Amersham Hypercassette™
Autoradiografiefilms Cassettes
Robuuste autoradiografiecassettes voor ontwikkelingen met zeer zwakke intensiteit en temperaturen tot –70 °C.
  • Robust and chip-proof autoradiography cassettes
  • Light-tight exposures at temperatures as low as –70 °C
  • Maximum contact between film and sample provides optimal resolution
  • Non porous pad gives a wipe-clean surface ensuring minimum contamination
Amersham Hypercassette autoradiography cassettes are constructed from strong aluminium and designed for the toughest demands of life science research. Deep cassettes are 1,5 cm in depth, allowing them to accommodate thicker samples. A choice of Hypercassette colors enables color coding of work. Use a permanent marker to record details on the polycarbonate label and be confident of secure identification, then wipe clean after use.
Bestelinformatie: Andere maten op verzoek leverbaar. Neem voor nadere informatie contact met ons op.