Sampling spoons, long handle, disposable, LaboPlast®/SteriPlast®
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
331-0326EA 18.9 EUR
331-0326 331-0325 331-0324 331-0323
Sampling spoons, long handle, disposable, LaboPlast®/SteriPlast®
Lepels
This sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
  • PS, white
  • Individually packed and optionally sterilised
  • Handle length: 300 mm
  • For single use
  • Clean room manufactured (clean room Class 7)
The long handle makes it easy to reach the bottoms of larger containers as well as sampling of drums, jerricans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand. The laboratory spoon is suitable for single-use sensitive areas in pharmaceuticals, chemistry, cosmetics, animal feed also food and beverage.
Certificaten: Complies with EU food requirements and FDA regulations.