VWR®, Accessories for Dissolved Oxygen Meter Set, Portable DO-230
Supplier: VWR Collection
Certificates
About this item
Product Information
- Catalog #PkAvailabilityPrice
- DO sensor DO OSG-1T, with a built-in temperature sensorDO sensor DO OSG-1T, with a built-in temperature sensor664-02371Log in for availability1 * 1 ST243,00 €Specifications:
Specifications
Environmentally Preferable - Maintenance set for the DO sensor MS-OSG-1, set includes: 3× spare membrane, 2×30 ml bottle of electrolyte, sandpaperMaintenance set for the DO sensor MS-OSG-1, set includes: 3× spare membrane, 2×30 ml bottle of electrolyte, sandpaper664-02381Log in for availability1 * 1 ST82,50 €Specifications:
Specifications
Environmentally Preferable