Netherlands
Order Entry
Autoclave type test packs, bowie and dick type, Comply™
  111-1033
undefined
Autoclave type test packs, bowie and dick type, Comply™
  111-1033
358,00 €
  1 * 20 ST
  • Type:
    Sterilization
  • Colour:
    Yellow
  • Description:
    Bowie and Dick type autoclave test pack, Comply™, disposable, colour turns from red to blue (1300)
  • Seq:
    01
  • Environmentally Preferable:
  • Fixture time:
    12 mins
  • Operating temperature:
    < 230 °C
  • Pk:
    20
The Bowie and Dick test is carried out daily in an empty chamber after a warm up cycle and is designed to show the adequacy of steam penetration.
  • Enables compliance with the HTM 01-05 as set by the Department of Health, requiring Dental practitioners to conduct a daily Bowie & Dick test in vacuum sterilizers
  • Test pack will give peace of mind and prove your sterilizer is working adequately
  • Saves time - the obvious colour change allows easy interpretation, simple and easy to use
  • Accurate, consistent, reliable and reproducible
  • Reduced storage space - compact and disposable
  • Cost saving - paper can be re-used for note-taking
  • Irreversible, for safe storage and traceability
  • One pack of 20 sheets will last a month - saving time on placing orders
Helping you to meet the requirements of the Dental sterilization standards as set by the Department of Health (HTM 01-05.) The Comply Bowie & Dick type test pack is designed to detect air leaks, inadequate air removal and steam penetration for vacuum-steam sterilizers.