Gowns, Tychem® 2000 C, model 0290
Supplier: DuPont
Certificates
About this item
DuPont™ Tychem® 2000 C gown model 0290 is shin-length gown with wrap-over rear closure, hook and loop neck closure and waist ties. Double cuffs for increased protection and tighter connection with gloves.
- Elasticated wrists
- Knitted cuff (inside) for better comfort
- Partial body chemical protective clothing
- Mandarin collar for improved neck and throat area closure
- Stitched and over-taped seams with barrier tape for protection and strength
Specially designed for use with Tychem® apparel,Tychem® Accessories can help offer enhanced protection for body parts that are more exposed to hazardous substances.
Tychem® 2000 C is used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
Certifications: Certified according to regulation (EU) 2016/425, category III, Type PB [3-B], EN 14126 (barrier to infective agents), antistatic treatment (EN 1149-1) - on inside.
Specifications
- Catalog No:
- 114-9331
- 114-9332
- 114-9333
- Gender:
- Unisex
- Unisex
- Unisex
Specifications
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: TC0290TYL00SMLocal Article Number: DUPOTC0290TYL00SMSpecifications:
More
Specifications
Environmentally PreferableGenderUnisexGarment application - Supplier Article Number: TC0290TYL00L2XLocal Article Number: DUPOTC0290TYL00L2XSpecifications:
More
Specifications
Environmentally PreferableGenderUnisexGarment application - Supplier Article Number: TC0290TYL0034XLocal Article Number: DUPOTC0290TYL0034XSpecifications:
More
Specifications
Environmentally PreferableGenderUnisexGarment application
Frequently Bought Together