5-Bromo-6-methylisoquinoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1146298-61-4
:
Formula:C₁₀H₈BrN
MDL Number:MFCD09264577
:222.081 g/mol
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR300110-100MGLocal Article Number: APOSOR300110-100MG
- Supplier Article Number: OR300110-250MGLocal Article Number: APOSOR300110-250MG