Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
1-(Bromomethyl)-3-methoxycyclobutane
1-(Bromomethyl)-3-methoxycyclobutane 95%
1-(Bromomethyl)-3-methoxycyclobutane 95%
1-(Bromomethyl)-3-methoxycyclobutane 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(Bromomethyl)-3-methoxycyclobutane 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1696252-78-4
:
Formula:
C₆H₁₁BrO
MDL Number:
MFCD30487495
:
179.055 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR55389-100MG
OR55389-100MG
Bottle
100 mg
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 681,00 €
681,00 €
-
+
Add to cart
1 * 100 mg
681,00 €
-
+
Add to cart
APOSOR55389-250MG
OR55389-250MG
Bottle
250 mg
Log in
for availability
Log in
for availability
1 * 250 mg
1 * 250 mg - 966,00 €
966,00 €
-
+
Add to cart
1 * 250 mg
966,00 €
-
+
Add to cart