2,2-Dimethyloxan-4-ol 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24945-13-9
:
Formula:C₇H₁₄O₂
MDL Number:MFCD01549685
:130.185 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR919422-1GLocal Article Number: APOSOR919422-1G
- Supplier Article Number: OR919422-5GLocal Article Number: APOSOR919422-5G