Sodium 1-cyano-3-ethoxy-2,3-dioxopropan-1-ide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:53544-13-1
:
Formula:C₆H₆NNaO₃
MDL Number:MFCD24696331
:163.107 g/mol
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR961901-250MGLocal Article Number: APOSOR961901-250MG
- Supplier Article Number: OR961901-1GLocal Article Number: APOSOR961901-1G