(2S)-3-Ethylazetidine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD29918540
:129.157 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR967369-250MGLocal Article Number: APOSOR967369-250MG
- Supplier Article Number: OR967369-500MGLocal Article Number: APOSOR967369-500MG
- Supplier Article Number: OR967369-1GLocal Article Number: APOSOR967369-1G