2-(2,5-Difluorophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetic acid
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₂₃H₁₇F₂NO₄
MDL Number:MFCD31420776
:409.382 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice