Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
2-(4-Trifluoromethyl-phenyl)-1H-imidazole
2-(4-Trifluoromethyl-phenyl)-1H-imidazole
2-(4-Trifluoromethyl-phenyl)-1H-imidazole
2-(4-Trifluoromethyl-phenyl)-1H-imidazole
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4-Trifluoromethyl-phenyl)-1H-imidazole
Supplier:
Apollo Scientific
About this item
CAS Number:
34898-30-1
:
Formula:
C₁₀H₇F₃N₂
MDL Number:
MFCD00106513
:
212.171 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSPC446033-1G
PC446033-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 184,00 €
184,00 €
-
+
Add to cart
1 * 1 g
184,00 €
-
+
Add to cart
APOSPC446033-5G
PC446033-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 902,00 €
902,00 €
-
+
Add to cart
1 * 5 g
902,00 €
-
+
Add to cart