Netherlands
Order Entry
3-(Difluoromethyl)-2-(2,3,4-trimethoxyphenyl)-1H-indole
3-(Difluoromethyl)-2-(2,3,4-trimethoxyphenyl)-1H-indole
Catalog # APOSPC520098-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethyl)-2-(2,3,4-trimethoxyphenyl)-1H-indole
Catalog # APOSPC520098-1G
Supplier:  Apollo Scientific
CAS Number:  
Supplier Article Number: PC520098-1G
Local Article Number: APOSPC520098-1G
Price
2 760,00 €
Log in for availability
Unit of Measure 1 * 1 g

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :333.329 g/mol
Storage Temperature:Ambient
CAS Number:2149597-56-6
Formula:C₁₈H₁₇F₂NO₃
MDL Number:MFCD29767581