Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
1-Iodo-2-trifluoromethoxyethane
1-Iodo-2-trifluoromethoxyethane 94%
1-Iodo-2-trifluoromethoxyethane 94%
1-Iodo-2-trifluoromethoxyethane 94%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Iodo-2-trifluoromethoxyethane 94%
Supplier:
Apollo Scientific
About this item
CAS Number:
1032065-38-5
:
Formula:
C₃H₄F₃IO
MDL Number:
MFCD21303949
:
239.963 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSPC520892-500MG
PC520892-500MG
Bottle
500 mg
Log in
for availability
Log in
for availability
1 * 500 mg
1 * 500 mg - 794,00 €
794,00 €
-
+
Add to cart
1 * 500 mg
794,00 €
-
+
Add to cart
APOSPC520892-1G
PC520892-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 1 350,00 €
1 350,00 €
-
+
Add to cart
1 * 1 g
1 350,00 €
-
+
Add to cart