Netherlands
Order Entry
4-(2,4-Difluorophenyl)but-3-enoic acid 95%
4-(2,4-Difluorophenyl)but-3-enoic acid 95%
Supplier:  Apollo Scientific
Certificates
4-(2,4-Difluorophenyl)but-3-enoic acid 95%
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSPC907959-250MG
    PC907959-250MG
    Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 271,00 €
    271,00 €
    1 * 250 mg
       271,00 €
    Supplier Article Number: PC907959-250MG
    Local Article Number: APOSPC907959-250MG
  • APOSPC907959-1G
    PC907959-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 687,00 €
    687,00 €
    1 * 1 g
       687,00 €
    Supplier Article Number: PC907959-1G
    Local Article Number: APOSPC907959-1G