Bismuth(III) oxide 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1304-76-3
:
Formula:BiO₃
MDL Number:MFCD00003462
:465.96 g/mol
Boiling Pt:1890 °C
Melting Pt:817 °C
Density:8.9 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: IN1252-100GLocal Article Number: APOSIN1252-100G
- Supplier Article Number: IN1252-500GLocal Article Number: APOSIN1252-500G