Iron(II) sulphide 99.99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1317-37-9
:
Formula:FeS
MDL Number:MFCD00011013
:87.91 g/mol
Melting Pt:1195 °C
Density:4.74 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
:GHS07
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: IN2092-5GLocal Article Number: APOSIN2092-5G
- Supplier Article Number: IN2092-25GLocal Article Number: APOSIN2092-25G