Methyl-4,5-dimethoxy-2-nitrobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26791-93-5
:
Formula:C₁₀H₁₁NO₆
MDL Number:MFCD00007239
:241.2 g/mol
Melting Pt:141…144 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR0619-50GLocal Article Number: APOSOR0619-50G
- Supplier Article Number: OR0619-100GLocal Article Number: APOSOR0619-100G
- Supplier Article Number: OR0619-250GLocal Article Number: APOSOR0619-250G