2',4'-Dimethylacetanilide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2050-43-3
:
Formula:C₁₀H₁₃NO
MDL Number:MFCD00026125
:163.22 g/mol
Melting Pt:129…131 °C
EINECS:218-088-6
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR110433-5GLocal Article Number: APOSOR110433-5G
- Supplier Article Number: OR110433-25GLocal Article Number: APOSOR110433-25G